Organic reference standards, (+)-gamma-Tocopherol, analytical standard, Supelco®
47785.
: Merck
:
- Pk:1
- CAS No.:54-28-4
- Description:(+)-gamma-Tocopherol
- Formula:C₂₈H₄₈O₂
- Storage temperature:−20 °C
- Description:(+)-γ-Tocopherol
- Pk Info:25 mg/ampule
- Form:Neat