Supelco® Organic Reference Standard, (−)-Huperzine A
Catalog # 42643-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:102518-79-6
- Description:(−)-Huperzine A
- Storage conditions:2...8 °C
- Hazard class description:H300 + H310 + H330
- Shipping conditions:STD
- MDL No.:MFCD01714949
- M-Clarity quality level:MQ100
- Synonym(s):(−)-Selagine; Kimpukan A
- Formula:C₁₅H₁₈N₂O
- Molecular weight:242,32
- Storage temperature:2…8 °C
- Flash point:Not applicable
- Form:Neat
Specifications
About this item
- GHS signal: GHS06
Caution: P262 - P280 - P301 + P310 + P330 - P302 + P352 + P310 - P304 + P340 + P310