Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, (−)-Fenchone
Supelco® Organic Reference Standard, (−)-Fenchone
Catalog # 92101-50MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, (−)-Fenchone
Catalog # 92101-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    7787-20-4
  • Description:
    (−)-Fenchone
  • Storage conditions:
    RT
  • Hazard class description:
    H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00151104
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (1R)-(−)-Fenchone; (−)-1,3,3-Trimethyl-2-norbornanone; (−)-Fenchone; (1R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one
  • Melting point:
    5 - 6 °C (lit.)
  • Molecular weight:
    152,23
  • Density:
    0,948 g/ml at 25 °C (lit.)
  • Flash point:
    52 °C - Closed cup
  • Boiling point:
    192 - 194 °C (lit.)

Specifications

About this item

  • GHS signal: GHS02
Caution: P273 - P391 - P501