Supelco® Organic Reference Standard, (−)-Epicatechin
Catalog # 68097-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:490-46-0
- Description:(−)-Epicatechin
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:WICE
- MDL No.:MFCD00075648
- M-Clarity quality level:MQ100
- Synonym(s):(−)-cis-3,3′,4′,5,7-Pentahydroxyflavane; (2R,3R)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol
- Melting point:240 °C (dec.) (lit.)
- Molecular weight:290,27
Specifications
About this item
- GHS signal: GHS07
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338