Lucifer yellow CH dipotassium salt fluorescent dye, for histology
Catalog # 80016.
Supplier: Biotium
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
- Environmentally Preferable:
Specifications
About this item
Synonyms: LY CH potassium salt
:
:521.57 g/mol
Storage Temperature:Refrigerator
CAS Number:71206-95-6
Formula:C₁₃H₉K₂N₅O₉S₂
MDL Number:MFCD00013445