8-Aminonaphthalene-1,3,6-trisulphonic acid disodium salt fluorescent dye
Catalog # 90010.
Supplier: Biotium
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:427.32 g/mol
Storage Temperature:Refrigerator
CAS Number:5398-34-5
Formula:C₁₀H₉NO₉S₃
MDL Number:MFCD00070572