Order Entry
Italy
ContactUsLinkComponent
Di-8-ANEPPQ membrane stain, for histology
Di-8-ANEPPQ membrane stain, for histology
Catalog # 61014.
Supplier:  Biotium
CAS Number:  
undefined
Di-8-ANEPPQ membrane stain, for histology
Catalog # 61014.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :717.70 g/mol
Storage Temperature:Freezer
Formula:C₃₈H₅₉Br₂N₃