Order Entry
Italy
ContactUsLinkComponent
 
DiBAC₄(3) membrane stain, for histology
Catalog # 61011.
Supplier:  Biotium
CAS Number:  
undefined
DiBAC₄(3) membrane stain, for histology
Catalog # 61011.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Bis(1,3-dibutylbarbituric acid) trimethine oxonol, DiBAC4(3)
 :
 :516.63 g/mol
Storage Temperature:Freezer
CAS Number:70363-83-6
Formula:C₂₇H₄₀N₄O₆
MDL Number:MFCD07437857