Supelco® Organic Reference Standard, ASTM® D6584 Tricaprin Solution
CRM44897
: Merck
:
- Concentration:8000 µg/ml in pyridine
- Pk:5 mL
- Standards:Certified reference material
- CAS No.:621-71-6
- Description:ASTM® D6584 Tricaprin Solution
- Storage conditions:2…8 °C
- Hazard class description:H225 - H302 + H312 + H332 - H315 - H319
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Synonym(s):Biodiesel impurity
- Flash point:17,0 °C - Closed cup
- GHS signal: GHS02, GHS07
: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338