Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, all-cis-4, 7, 10, 13, 16, 19-Docosahexaenoic acid methyl ester
Supelco® Organic Reference Standard, all-cis-4, 7, 10, 13, 16, 19-Docosahexaenoic acid methyl ester
Catalog # CRM47570
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, all-cis-4, 7, 10, 13, 16, 19-Docosahexaenoic acid methyl ester
Catalog # CRM47570
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 mg/ml in heptane
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    all-cis-4, 7, 10, 13, 16, 19-Docosahexaenoic acid methyl ester
  • Storage conditions:
    –10...–25 °C
  • Hazard class description:
    H225 - H304 - H315 - H336 - H410
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Flash point:
    –3,99 °C – Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331