Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, EN 14105/D6584 Monoglyceride Stock Solution
Supelco® Organic Reference Standard, EN 14105/D6584 Monoglyceride Stock Solution
Catalog # 49446-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, EN 14105/D6584 Monoglyceride Stock Solution
Catalog # 49446-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 mg/ml each component in pyridine
  • Pk:
    1
  • Standards:
    Analytical standard
  • Description:
    EN 14105/D6584 Monoglyceride stock solution
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H315 - H319
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Biodiesel impurity; Commercial Mixture of Monoglycerides; Monoglyceride mix; Monoglyceride mixture; Monoglyceride standard; Monoglycerides

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338