Order Entry
Italy
ContactUsLinkComponent
Tetramethylammonium sulphate ≥99.0% (by titrimetric analysis), powder, LiChropur™ for ion pair chromatography, Supelco®
Irritant
Tetramethylammonium sulphate ≥99.0% (by titrimetric analysis), powder, LiChropur™ for ion pair chromatography, Supelco®
Catalog # 02799-10G
Supplier:  Merck
CAS Number:  
undefined
Irritant
Tetramethylammonium sulphate ≥99.0% (by titrimetric analysis), powder, LiChropur™ for ion pair chromatography, Supelco®
Catalog # 02799-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance (Color):
    Colorless or White
  • Titration (Ion Exchange):
    99.0 - 101.0 %
  • Suitability Test (HPLC):
    RP-Gradient Test Corresponds to Standard
  • Proton NMR Spectrum:
    Conforms to Structure
  • Test By UV Spectroscopy:
    210nm≤ 0.1
  • Cyclic Voltammogram:
    Corresponds to Standard CV
  • Residue (Filter Test):
    No Residue
  • Filtertest (Method):
    3g in 30ml Water (10%)
  • Recommended Retest Period:
    42 Months

Specification Test Results

About this item

 :
 :244.35 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:238-043-4
 :GHS07
CAS Number:14190-16-0
Formula:C₈H₂₄N₂O₄S
MDL Number:MFCD00012139