N-Methyl-bis-heptafluorobutyramide (MBHFBA) ≥96.0% (by GC), LiChropur™ for GC derivatisation, Supelco®
Catalog # 78268-1ML-F
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:423.10 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:73980-71-9
Formula:C₉H₃F₁₄NO₂
MDL Number:MFCD00049280