

N-Methyl-bis-heptafluorobutyramide (MBHFBA) acylation reagent, for GC derivatisation
Supplier: MACHEREY-NAGEL
Certificates
About this item
CAS Number:73980-71-9
:
Formula:C₉H₃F₁₄NO₂
MDL Number:MFCD00049280
:423.10 g/mol
Boiling Pt:165…166 °C (760 mm Hg)
Density:1.673 g/cm³ (d20°/4°)
:GHS02
:GHS07
Frequently Bought Together