Supelco® Organic Reference Standard, F.A.M.E. Mix Glc-30
Catalog # 1893-1AMP
Nome del fornitore: Merck
Numero CAS:
Specifiche
- Pk:1 Ampoul
- Standards:Certified reference material, ampule of 100 mg (Neat)
- Description:F.A.M.E. Mix GLC-30
- Storage conditions:–10...–25 °C
- Hazard class description:H410
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
- Synonym(s):Fatty Acid Methyl Ester Mix
- Pk Info:Ampoule of 100 mg
Specifiche
Informazioni su questo articolo
- GHS signal: GHS07
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Packaging: 100 mg ampule contains approximately equal amounts by weight of: Caprylic Acid Methyl Ester (C8:0) Capric Acid Methyl Ester (C10:0) Lauric Acid Methyl Ester (C12:0) Myristic Acid Methyl Ester (C14:0) Palmitic Acid Methyl Ester (C16:0)
Caution: P273 - P391 - P501