Supelco® Organic Reference Standard, F.A.M.E. Mix
Catalog # 18916-1AMP
Supplier: Merck
CAS Number:
Specifications
- Pk:1 Ampoul
- Standards:C18:0 - C20:0, Certified reference material
- Description:F.A.M.E. Mix
- Storage conditions:–20 °C
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
- Synonym(s):Fatty Acid Methyl Ester Mix
- Pk Info:Pkg of 100 mg
Specifications
About this item
Packaging: 100 mg ampule contains the following with the weight percentages as indicated. Stearic acid methyl ester (C18:0) 10% Oleic acid methyl ester (C18:1cis-9) 20% Elaidic acid methyl ester (C18:1trans-9) 20% Linoleic acid methyl ester (C18:2cis-912) 20%
Linolelaidic acid methyl ester (C18:2trans-912) 20%
Arachidic acid methyl ester (C20:0) 10%
Linolelaidic acid methyl ester (C18:2trans-912) 20%
Arachidic acid methyl ester (C20:0) 10%