Supelco® PESTANAL® Organic Reference Standard, Thionazin
Catalog # 33941-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:297-97-2
- Description:Thionazin
- Storage conditions:2...8 °C
- Hazard class description:H300 + H310
- Shipping conditions:STD
- MDL No.:MFCD00055349
- M-Clarity quality level:MQ100
- Synonym(s):O,O-Diethyl O-(2-pyrazinyl) phosphorothioate
- Formula:C₈H₁₃N₂O₃PS
- Molecular weight:248,24
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06
Caution: P262 - P264 - P270 - P280 - P301 + P310 - P302 + P352 + P310