Order Entry
Italy
ContactUsLinkComponent
Pesticide reference standard, Pyributicarb, analytical standard, Supelco®
Pesticide reference standard, Pyributicarb, analytical standard, Supelco®
Catalog # 92842-100MG
Supplier:  Merck
CAS Number:  
Pesticide reference standard, Pyributicarb, analytical standard, Supelco®
Catalog # 92842-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    88678-67-5
  • Description:
    Pyributicarb
  • Formula:
    C₁₈H₂₂N₂O₂S
  • Description:
    Pyributicarb

Specifications

About this item

Packaging: Bottomless glass bottles are inserted with fused cone.