Order Entry
Italy
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Prosulfocarb
Supelco® PESTANAL® Organic Reference Standard, Prosulfocarb
  31141-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Prosulfocarb
  31141-250MG
 :  Merck
 :  

 

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    52888-80-9
  • Description:
    Prosulfocarb
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H317 - H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00145179
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    S-Benzyl dipropylthiocarbamate
  • Formula:
    C₁₄H₂₁NOS
  • Molecular weight:
    251,39
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS06, GHS09
 : P261 - P264 - P273 - P280 - P301 + P312 - P302 + P352