Order Entry
Italy
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Perfluorotetradecanoic acid
  80312-50MG
 :  Merck
 :  
Supelco® Organic Reference Standard, Perfluorotetradecanoic acid
  80312-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    376-06-7
  • Description:
    Perfluorotetradecanoic acid
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 + H332 - H351 - H360D - H362 - H372
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00153263
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    PFAS
  • Melting point:
    130 - 135 °C (lit.)
  • Formula:
    CF₃(CF₂)₁₂CO₂H
  • Molecular weight:
    714,11
  • Boiling point:
    270 °C/740 mmHg (lit.)

 

 

  • GHS signal: GHS05
 : P202 - P260 - P263 - P301 + P312 - P304 + P340 + P312 - P308 + P313