Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, Methyl Elaidate
Supelco® Organic Reference Standard, Methyl Elaidate
Catalog # 45119-5ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Methyl Elaidate
Catalog # 45119-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1937-62-8
  • Description:
    Methyl elaidate
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00066521
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Elaidic acid methyl ester; Methyl trans-9-octadecenoate
  • Melting point:
    9...10 °C
  • Formula:
    CH₃(CH₂)₇CH=CH(CH₂)₇COOCH₃
  • Molecular weight:
    296,49
  • Storage temperature:
    2…8 °C
  • Density:
    0,871 g/ml at 20 °C (lit.)
  • Flash point:
    Not applicable

Specifications

About this item