Supelco® Organic Reference Standard, Simazine
: Merck
- GHS signal: GHS02
- CAS No.:122-34-9
- Storage conditions:RT
- Shipping conditions:STD
- MDL No.:MFCD00023174
- M-Clarity quality level:MQ100
- Synonym(s):2,4-Bis(ethylamino)-6-chloro-1,3,5-triazine
- Formula:C₂₃H₄₄O₂
- Molecular weight:201,66
- Density:0,870 g/ml at 20 °C (lit.)
- Flash point:33,0 °C - closed cup
- Boiling point:172...175 °C/0,2 mm Hg (lit.)
Frequently Bought Together