Supelco® PESTANAL® Organic Reference Standard, Glyphosate-2-¹³C
Catalog # 90007-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:287399-31-9
- Description:Glyphosate-2-¹³C
- Storage conditions:RT
- Hazard class description:H318 - H411
- Shipping conditions:STD
- MDL No.:MFCD01075599
- M-Clarity quality level:MQ200
- Synonym(s):N-(Phosphonomethyl)glycine-2-¹³C
- Melting point:230 °C (dec.) (lit.)
- Formula:HO₂C¹³CH₂NHCH₂PO(OH)₂
- Molecular weight:170,07
Specifications
About this item
- GHS signal: GHS05, GHS09
Caution: P280 - P305 + P351 + P338 + P310