Supelco® PESTANAL® Organic Reference Standard, Flonicamid Metabolite Tfng
Catalog # 34566-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:207502-65-6
- Description:Flonicamid Metabolite TFNG
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD11112215
- M-Clarity quality level:MQ100
- Synonym(s):N-[[4-(Trifluoromethyl)-3-pyridinyl]carbonyl]glycine; 4-(Trifluoromethyl)nicotinoyl glycine
- Formula:C₉H₇F₃N₂O₃
- Molecular weight:248,16
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Caution: P301 + P312 + P330