Supelco® PESTANAL® Organic Reference Standard, Edifenphos
Catalog # 45467-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:17109-49-8
- Description:Edifenphos
- Storage conditions:2...8 °C
- Hazard class description:H301 + H311 + H331 - H317 - H410
- Shipping conditions:STD
- MDL No.:MFCD00078673
- M-Clarity quality level:MQ100
- Synonym(s):O-Ethyl S,S-diphenyl phosphorodithioate
- Formula:C₁₄H₁₅O₂PS₂
- Molecular weight:310,37
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P261 - P273 - P280 - P301 + P310 - P302 + P352 + P312 - P304 + P340 + P311