Supelco® Organic Reference Standard, Conjugated (9E,11E)-Linoleic acid
Catalog # 90983-20MG
Supplier: Merck
CAS Number:
Specifications
- Pk:20 mg
- Standards:Analytical standard
- CAS No.:544-71-8
- Description:Conjugated (9E,11E)-Linoleic acid
- Storage conditions:–20 °C
- Shipping conditions:DICE
- MDL No.:MFCD00133681
- M-Clarity quality level:MQ100
- Synonym(s):(9E,11E)-9,11-Octadecadienoic acid; 9E,11E-CLA; Isolinoleic acid; Linoleic acid (9-trans, 11-trans); Mangold’s acid
- Molecular weight:280,45