Order Entry
Italy
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Bixafen
Supelco® PESTANAL® Organic Reference Standard, Bixafen
Catalog # 32581-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Bixafen
Catalog # 32581-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    581809-46-3
  • Description:
    Bixafen
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H400
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD16652583
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N-(3′,4′-Dichloro-5-fluorobiphenyl-2-yl)-3-(difluoromethyl)-1-methylpyrazole-4-carboxamide
  • Formula:
    C₁₈H₁₂Cl₂F₃N₃O
  • Molecular weight:
    414,21
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS09
Caution: P273 - P391 - P501