Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Bisoctrizole, analytical standard, Supelco®
Organic reference standard, Bisoctrizole, analytical standard, Supelco®
Catalog # 30184-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Bisoctrizole, analytical standard, Supelco®
Catalog # 30184-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    103597-45-1
  • Description:
    Bisoctrizole
  • Melting point:
    197...199 °C (lit.)
  • Formula:
    C₄₁H₅₀N₆O₂
  • Description:
    Bisoctrizole

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.