Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Bemotrizinol, analytical standard, Supelco®
Organic reference standard, Bemotrizinol, analytical standard, Supelco®
Catalog # 51825-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Bemotrizinol, analytical standard, Supelco®
Catalog # 51825-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    187393-00-6
  • Description:
    Bemotrizinol
  • Melting point:
    85...87 °C
  • Formula:
    C₃₈H₄₉N₃O₅
  • Description:
    Bemotrizinol

Specifications

About this item