Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, BDE No 77 Solution
Supelco® Organic Reference Standard, BDE No 77 Solution
Catalog # 34115-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, BDE No 77 Solution
Catalog # 34115-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    93703-48-1
  • Description:
    BDE No 77 solution
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H304 - H315 - H336 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00626455
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3,3′,4,4′-Tetrabromodiphenyl ether solution; 3,3′,4,4′-TetraBDE; PBDE 77
  • Formula:
    C₁₂H₆Br₄O
  • Molecular weight:
    485,79
  • Flash point:
    –12 °C - closed cup

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P273 - P301 + P310 + P331 - P302 + P352