Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, BDE No 28
Supelco® Organic Reference Standard, BDE No 28
Catalog # 05364-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, BDE No 28
Catalog # 05364-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    41318-75-6
  • Description:
    BDE No 28
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD09037597
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2,4,4′-TriBDE; 2,4,4′-Tribromodiphenyl ether; PBDE 28
  • Melting point:
    63...66 °C
  • Formula:
    C₁₂H₇Br₃O
  • Molecular weight:
    406,90
  • Flash point:
    Not applicable

Specifications

About this item