Supelco® PESTANAL® Organic Reference Standard, 2-Isopropylthioxanthone
Catalog # 34221-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:5495-84-1
- Description:2-Isopropylthioxanthone
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00055658
- M-Clarity quality level:MQ100
- Synonym(s):2-Isopropyl-9H-thioxanthen-9-one
- Formula:C₁₆H₁₄OS
- Molecular weight:254,35
- Flash point:Not applicable