VISOCOLOR® Colorimetric Test, Nickel
Supplier: MACHEREY-NAGEL
Certificates
About this item
Reagents for the determination of nickel in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Remark: Measuring range on PF-12Plus. Range on other photometers can be different.
Hazardous material: Yes
Specifications
- Model:MANA931240
- Platform:Colorimetric test kits
- Storage temperature:15...25 °C
- Shelf life (months):18
- Weight:144,3 g (Including packaging)