Thiamine hydrochloride, powder
:67-03-8
:
:C₁₂H₁₈Cl₂N₄OS
:MFCD00012780
:337.27 g/mol
:164 °C
:Ambient
:200-641-8
- Identity Test:Passes
- FTIR:Conforms to Standard
- pH:2,7 - 3,4 (1% aq soln)
- UV/VIS: OD400:≤0,25 (1% aq soln)
- Loss on Drying:≤5,0%
- Residue on Ignition:≤0,2%
Frequently Bought Together