2-Oleoyl Glycerol solution ≥98% in ethanol
Supplier: Cayman Chemical
Certificates
About this item
Synonyms: 2-Monoolein, 2-Hydroxy-1-(hydroxymethyl)ethyl ester-9Z-octadecenoic acid, 2-OG
CAS Number:3443-84-3
:
Formula:C₂₁H₄₀O₄
MDL Number:MFCD00171536
:356.5 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
UN:1170
ADR:3,II
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity:≥98%
- Formulation:A Solution in Ethanol
Specification Test Results
Frequently Bought Together