Order Entry
Italy
ContactUsLinkComponent
 
Pharmaceutical analytical standard, Quinoxaline-2-carboxylic acid methylester, reference material, Honeywell Fluka™
  HONCR1475-10MG
 :  
Pharmaceutical analytical standard, Quinoxaline-2-carboxylic acid methylester, reference material, Honeywell Fluka™
  HONCR1475-10MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    1865-11-8
  • Description:
    Quinoxaline-2-carboxylic acid methylester
  • Formula:
    C₁₀H₈N₂O₂
  • Storage temperature:
    Ambient
  • Description:
    Quinoxaline-2-carboxylic acid methylester
  • Freezing point:
    112...113 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results