Order Entry
Italy
ContactUsLinkComponent
Pharmaceutical analytical standard, 5-Hydroxyflunixin-d3, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, 5-Hydroxyflunixin-d3, reference material, Honeywell Fluka™
Catalog # HONCR1456-10MG
CAS Number:  
Pharmaceutical analytical standard, 5-Hydroxyflunixin-d3, reference material, Honeywell Fluka™
Catalog # HONCR1456-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    1185088-54-3
  • Description:
    5-Hydroxyflunixin-d₃
  • Formula:
    C₁₄H₈D₃F₃N₂O₃
  • Storage temperature:
    Ambient
  • Description:
    5-Hydroxyflunixin-d₃
  • Freezing point:
    299...306 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results