Pharmaceutical analytical standard, (±)-Propranolol-(isopropyl-d7) hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1161-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1613439-56-7
- Description:(±)-Propranolol-(isopropyl-d₇) hydrochloride
- Formula:C₁₆D₇H₁₄NO₂ · HCl
- Storage temperature:2...8 °C
- Description:(±)-Propranolol-(isopropyl-d₇) hydrochloride
- Freezing point:160 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results