Pharmaceutical analytical standard, 4-Acetamido-d3-antipyrine, reference material, Honeywell Fluka™
Catalog # HONCR1159-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:342821-66-3
- Description:4-Acetamido-d₃-antipyrine
- Formula:C₁₃D₃H₁₂N₃O₂
- Storage temperature:2...8 °C
- Description:4-Acetamido-d₃-antipyrine
- Freezing point:201 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results