Order Entry
Italy
ContactUsLinkComponent
 
Pesticide reference standard, Dimefluthrin, neat, Honeywell Fluka™
  HONCR1125-100MG
 :  
Pesticide reference standard, Dimefluthrin, neat, Honeywell Fluka™
  HONCR1125-100MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • CAS No.:
    271241-14-6
  • Description:
    Dimefluthrin
  • Formula:
    C₁₉H₂₂F₄O₃
  • Storage temperature:
    2...8 °C
  • Description:
    Dimefluthrin
  • Flash point:
    161 °C
  • Freezing point:
    352 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results