Order Entry
Italy
ContactUsLinkComponent
Pharmaceutical analytical standard, Butoconazole, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Butoconazole, reference material, Honeywell Fluka™
Catalog # HONCR1120-100MG
Nome del fornitore:  Honeywell Chemicals
Numero CAS:  
Pharmaceutical analytical standard, Butoconazole, reference material, Honeywell Fluka™
Catalog # HONCR1120-100MG
Nome del fornitore:  Honeywell Chemicals
Numero CAS:  

Specifiche

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • CAS No.:
    64872-76-0
  • Description:
    Butoconazole
  • Formula:
    C₁₉H₁₇Cl₃N₂S
  • Storage temperature:
    Ambient
  • Description:
    Butoconazole
  • Freezing point:
    78 °C

Specifiche

Informazioni su questo articolo

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results