Order Entry
Italy
ContactUsLinkComponent
 
Sample vials, KIMBLE®
Catalog # SCERI50862300C4K2
Supplier:  DWK Life Sciences
undefined
Sample vials, KIMBLE®
Catalog # SCERI50862300C4K2
Supplier:  DWK Life Sciences

Specifications

  • Cap included:
    No
  • Cap Size:
    20-400
  • Ø×H:
    23×86 mm
  • Capacity:
    24,00 ml
  • Colour:
    Clear
  • Pk:
    200

Specifications

About this item