Imidazole, EMPROVE® EXPERT ACS
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:288-32-4
:
Formula:C₃H₄N₂
MDL Number:MFCD00005183
Boiling Pt:256 °C (1013 hPa)
Melting Pt:90 °C
Density:1.233 g/cm³ (20 °C)
Flash Pt:145 °C
UN:3263
ADR:8,
:68.08 g/mol
:GHS05
:GHS08
:GHS07
Frequently Bought Together