Sodium bromide ≥99%, Reagent Grade
Drum with inliner, 12 kg
Supplier: Honeywell Chemicals
Sodium bromide ≥99%, Reagent Grade
Catalog # 220345-12KG
Supplier: Honeywell Chemicals
CAS Number: 7647-15-6
Jump to:
Product Details & Documents
- ≥95% for general laboratory use.
- CAS Number:7647-15-6
- Formula:NaBr
- MDL Number:MFCD00003475
- Molecular Weight:102.89 g/mol
- Merck Index:13,08666
- Boiling Pt:1.393 °C (1.013 hPa)
- Melting Pt:750 °C
- Density:3.2 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:231-599-9
Specifications
Specification Test Results
- Assay:99.0 - 101.0 %
- Assay of Br:76.8 - 78.4 %
Specifications
- Pack type:Drum with inliner
- Pk:12 kg



