Order Entry
Italy
ContactUsLinkComponent
Potassium bromide 0.1 M KBr volumetric solution, Fluka™
Potassium bromide 0.1 M KBr volumetric solution, Fluka™
Catalog # 34257-1L
CAS Number:  
Potassium bromide 0.1 M KBr volumetric solution, Fluka™
Catalog # 34257-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :119 g/mol
Storage Temperature:Ambient
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358