Iron(II) sulphate concentrate for dilution to 1L, 0.1 M FeSO4 volumetric solution, Fluka™
Catalog # 38047-1EA
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:1
- Seq:0001
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:151.91 g/mol
Density:1.300 g/cm³ (20 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:7720-78-7
Formula:FeO₄S
MDL Number:MFCD00011014