Order Entry
Italy
ContactUsLinkComponent
EDTA magnesium dipotassium salt ≥98% for complexometry, Fluka™
EDTA magnesium dipotassium salt ≥98% for complexometry, Fluka™
Certificates
EDTA magnesium dipotassium salt ≥98% for complexometry, Fluka™

Specification Test Results

  • Assay:
    Min. 98 %
  • pH (0.1 M, 20°C):
    8.5 - 9.5
  • Calcium (Ca):
    Max. 0.05 %
  • Iron (Fe):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 0.01 %

Specification Test Results

Frequently Bought Together