Calcium chloride dihydrate ≥99.0%, Reagent Grade
:10035-04-8
:
:CaCl₂·2H₂O
:MFCD00149613
:147.01 g/mol
:1.900 °C (1.013 hPa)
:765 °C
:Ambient
:233-140-8
:GHS07
- Assay:99.0 - 105.0 %
- Appearance of the solution:complying
- Colour of the solution:complying
Frequently Bought Together