Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, C7 - C40 Saturated Alkanes Standard
Supelco® Organic Reference Standard, C7 - C40 Saturated Alkanes Standard
Catalog # 49452-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, C7 - C40 Saturated Alkanes Standard
Catalog # 49452-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml each component in hexane
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    C7 - C40 saturated alkanes standard
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H304 - H315 - H336 - H361f - H373 - H411
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    –26 °C – Closed cup

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07, GHS09
Certifications: Certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Caution: P202 - P210 - P273 - P301 + P310 - P303 + P361 + P353 - P331