Order Entry
Italy
ContactUsLinkComponent
Bromide bromate 0.25 M Br2 (0.5 N) volumetric solution, Fluka™
Health
Bromide bromate 0.25 M Br2 (0.5 N) volumetric solution, Fluka™
Catalog # 35007-1L
CAS Number:  
Health
Bromide bromate 0.25 M Br2 (0.5 N) volumetric solution, Fluka™
Catalog # 35007-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
Density:1.05 g/cm³
 :GHS08
Storage Temperature:Ambient
 :7758-01-2
Formula:KBrO₃/KBr